Learn More
IMEB, Inc Histo-Vacuum with 1 Disposable Hose and Hepa Filter
SDP
Supplier:  IMEB, Inc IHV
Encompass
IMEB Histo Vacuum; includes 1 disposable hose and disposable hepa filter. Fast and easy cleanup for microtomes and cryostats! Convenient shoulder strap. 4.0 amp motor.
Catalog No. NC0947502