Learn More
Diagnostic Biosystems HistoZyme
Catalog No. 502407589
Encompass_Preferred
HistoZyme is a proteolytic enzyme solution that results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, HistoZyme requires only a brief incubation at room temperature. Performing antigen recovery with HistoZyme, will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.\n\nFormalin fixation of tissues induces protein cross links that helps in maintaining the cellular morphology by inactivating the digestive enzymes and preserving the cytoskeleton. Fixation stops tissue autolysis, preserves tissue structures, and immobilizes antigens. However, antigens undergo chemical alteration of their primary, secondary and tertiary structures during fixation.
Catalog No. 50-240-7589 Supplier Diagnostic Biosystems Supplier No. K04650
HistoZyme is a proteolytic enzyme solution that results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, HistoZyme requires only a brief incubation at room temperature. Performing antigen recovery with HistoZyme, will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.\n\nFormalin fixation of tissues induces protein cross links that helps in maintaining the cellular morphology by inactivating the digestive enzymes and preserving the cytoskeleton. Fixation stops tissue autolysis, preserves tissue structures, and immobilizes antigens. However, antigens undergo chemical alteration of their primary, secondary and tertiary structures during fixation.