Learn More
Electron Microscopy Sciences Histobond Histobond + Supa Mega Slides
SDP
Catalog No. 501928716
Encompass_Preferred
Histobond + Supa Mega Slides are designed for mounting large sections of neuro and breast tissue. Manufactured from the highest quality white glass, with an innovative positive charge coating, they offer exceptional adhesive properties and provide a superior coverage of reagents across the slide.
These slides have a 'Right Side' indicator (RS) to help ensure users correctly orientate the slide, as well as a 7mm marking area across the width of the slide in order to maximize the working area.
Size: 75.2 x 50.4 1 mm
Catalog No. 50-192-8716 Supplier Electron Microscopy Sciences Supplier No. 7188160
Histobond + Supa Mega Slides are designed for mounting large sections of neuro and breast tissue. Manufactured from the highest quality white glass, with an innovative positive charge coating, they offer exceptional adhesive properties and provide a superior coverage of reagents across the slide.
These slides have a 'Right Side' indicator (RS) to help ensure users correctly orientate the slide, as well as a 7mm marking area across the width of the slide in order to maximize the working area.
Size: 75.2 x 50.4 1 mm