Learn More
HistoChoice™ Clearing Agent, Sigma-Aldrich
Catalog No. 01258962 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
01-258-962 1 L
1 options
Catalog No. 01-258-962 Supplier MilliporeSigma Supelco Supplier No. H27791L
Add to Cart
Add to Cart
HistoChoice™ Clearing Agent has the following applications: It is suitable as an alternative to toluene and xylene for dewaxing paraffin tissue sections. Prepared slides are suitable for antibody probing applications and in situ hybridizations. It has been employed in a study to explain the recalcitrance of lignocellulosic plant cell walls to enzymatic degradation. It has been used to develop a novel method to improve sensitivity for the localization of miRNA and their target transcripts in plants. It has been used to study the correlation of cytoplasmic ADP-ribosylation levels with markers of patient outcome in human cancers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Xylene Substitute
Quantity 1 L
Form Liquid