Learn More
Bioworld HISTOCHOICE MB TISSUE FIXATIVE
Catalog No. 50199053
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. HistoChoice MB Tissue Fixative The Molecular Biology Fixative Safe - Contains no formaldehyde, glutaraldehyde or mercury. Non-Toxic - Is completely odorless and can be safely disposed of down the drain. HISTOCHOICE MB Tissue Fixativ...
Catalog No. 50-199-053 Supplier Bioworld Supplier No. 217300081
This item has a minimum order quantity of 2 per supplier requirements. HistoChoice MB Tissue Fixative The Molecular Biology Fixative Safe - Contains no formaldehyde, glutaraldehyde or mercury. Non-Toxic - Is completely odorless and can be safely disposed of down the drain. HISTOCHOICE MB Tissue Fixativ...