Learn More
Sigma Aldrich Fine Chemicals Biosciences HistoChoiceR Clearing Agen1L
Catalog No. 502900116
Encompass
HistoChoice(R) is a patented complex mixture of agents that allow tissues to retain their structure antigenic sites and nucleic acid sites. This formulation in non-citrus contains no mercury formaldehyde or glutaraldehyde.We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry compared to the standard use of formalin for tissue processing.
Catalog No. 50-290-0116 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H27791L
HistoChoice(R) is a patented complex mixture of agents that allow tissues to retain their structure antigenic sites and nucleic acid sites. This formulation in non-citrus contains no mercury formaldehyde or glutaraldehyde.We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry compared to the standard use of formalin for tissue processing.