Learn More
StatLab™ HistoCool Tissue Cassette Cooling Tray
Description
- Trays are filled with freezer gel that stays frozen longer: up to 6 to 8 hours
- Simply refreeze at the end of each shift or day
- Red Tray has a 20 cassette capacity
- Blue Tray has a 30 cassette capacity
Specifications
Specifications
| Product Type | Tissue Cassette Drawer |
| Color | Red |