Learn More
StatLab™ HistoCool Tissue Cassette Cooling Tray
SDP
Catalog No. STLSLHC30
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLSLHC30 Blue
STLSLHC20 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STLSLHC30 Supplier StatLab™ Supplier No. SLHC30
Add to Cart
Add to Cart
Effectively aids in the cooling of tissue blocks.
  • Trays are filled with freezer gel that stays frozen longer: up to 6 to 8 hours
  • Simply refreeze at the end of each shift or day
  • Red Tray has a 20 cassette capacity
  • Blue Tray has a 30 cassette capacity
Specifications
Product Type Tissue Cassette Drawer
Color Blue