Learn More
Leica Biosystems Leica HistoCore BIOCUT Manual Rotary Microtome
Catalog No. NC2190273
Encompass
The HistoCore BIOCUT manual rotary microtome delivers precise, low-maintenance manual sectioning of formalin-fixed, paraffin-embedded tissue for routine histology and in vitro diagnostic workflows. It combines ergonomic controls, safety features, and easy cleaning to improve laboratory throughput.
  • Section thickness range 1-60 μm with selectable increments for fine and coarse cutting.
  • Two-in-one blade holder accommodates high and low profile blades for workflow flexibility.
  • Antistatic waste tray reduces paraffin adhesion and simplifies cleaning.
  • Personalized coarse feed wheel with user-selectable turn direction for ergonomic comfort.
  • Operator-adjustable force balancing and two handwheel locking systems for safe operation.
  • Silent specimen retraction and removable top tray for quiet, organized work areas.
This item is not returnable. View return policy
Catalog No. NC2190273 Supplier Leica Biosystems Supplier No. 149BIO000C1
Add to Cart
Add to Cart
Request a Quote
The HistoCore BIOCUT manual rotary microtome delivers precise, low-maintenance manual sectioning of formalin-fixed, paraffin-embedded tissue for routine histology and in vitro diagnostic workflows. It combines ergonomic controls, safety features, and easy cleaning to improve laboratory throughput.
  • Section thickness range 1-60 μm with selectable increments for fine and coarse cutting.
  • Two-in-one blade holder accommodates high and low profile blades for workflow flexibility.
  • Antistatic waste tray reduces paraffin adhesion and simplifies cleaning.
  • Personalized coarse feed wheel with user-selectable turn direction for ergonomic comfort.
  • Operator-adjustable force balancing and two handwheel locking systems for safe operation.
  • Silent specimen retraction and removable top tray for quiet, organized work areas.