Learn More
Leica Biosystems HistoCore Slide Dryer (Accessory for HistoCore Water Bath)
Catalog No. NC3665925
Encompass
The Leica Biosystems HistoCore Slide Dryer is a compact, space-saving accessory that accelerates drying of histology slides when used with a compatible water bath. It provides controlled heated air to speed up workflow and improve throughput in pathology and histology laboratories.
  • Space-saving design with small footprint.
  • Dries up to 30 slides.
  • Heated drying up to 75 °C for faster processing.
  • Compact dimensions: 200 x 280 x 98 mm.
  • Easy integration into standard histology slide-processing workflows.
This item is not returnable. View return policy
Catalog No. NC3665925 Supplier Leica Biosystems Supplier No. 140607010C0
The Leica Biosystems HistoCore Slide Dryer is a compact, space-saving accessory that accelerates drying of histology slides when used with a compatible water bath. It provides controlled heated air to speed up workflow and improve throughput in pathology and histology laboratories.
  • Space-saving design with small footprint.
  • Dries up to 30 slides.
  • Heated drying up to 75 °C for faster processing.
  • Compact dimensions: 200 x 280 x 98 mm.
  • Easy integration into standard histology slide-processing workflows.