Learn More
Leica Biosystems HistoCore SPECTRA H&E Stain System S1-800
Catalog No. NC3849065
Encompass
Leica Biosystems' HistoCore SPECTRA S1-800 is a ready-to-use H&E stain system designed for low-volume histology laboratories. It delivers reproducible H&E and special stain results across runs, reducing variability and turnaround time while supporting automated workflows.
  • Automated staining workflow with protocol initiation and tracking.
  • Consistent staining from slide 1 to 800.
  • Suitable for H&E and common special stains.
  • Reduces day-to-day variation and manual handling.
  • Formulated to deliver reliable, reproducible stain intensity.
Catalog No. NC3849065 Supplier Leica Biosystems Supplier No. 3801656
Leica Biosystems' HistoCore SPECTRA S1-800 is a ready-to-use H&E stain system designed for low-volume histology laboratories. It delivers reproducible H&E and special stain results across runs, reducing variability and turnaround time while supporting automated workflows.
  • Automated staining workflow with protocol initiation and tracking.
  • Consistent staining from slide 1 to 800.
  • Suitable for H&E and common special stains.
  • Reduces day-to-day variation and manual handling.
  • Formulated to deliver reliable, reproducible stain intensity.