Learn More
Electron Microscopy Sciences Histocryl Resin Kit
SDP
Supplier:  Electron Microscopy Sciences 14390
Encompass_Preferred
Histocryl acrylic resin has been specially formulated for light microscopy. It is water clear, and because it is hydrophilic (polar) it permits the use of most routine staining techniques without priorremoval or etching. Kit consists of: 500 ml Histocryl Resin , 50g Histocryl Catalyst (B.P.) , 10 ml Histocryl Accelerator
Catalog No. 50-980-426