Learn More
Electron Microscopy Sciences DiATOME Histo Cryo Diamond Knife Wet 4.0 mm 45° Knife angle
SDP
Catalog No. 501898846
Encompass
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission.
Size: 4.0 mm
Knife angle: 45°
Catalog No. 50-189-8846 Supplier Electron Microscopy Sciences Supplier No. 40HWC
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission.
Size: 4.0 mm
Knife angle: 45°