Learn More
Electron Microscopy Sciences DiATOME Histo Cryo Diamond Knife Wet 8.0 mm
SDP
Catalog No. 501929117
Encompass
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission.
Size: 8.0 mm
Knife angle: 45°
Catalog No. 50-192-9117 Supplier Electron Microscopy Sciences Supplier No. 80HWC
Add to Cart
Add to Cart
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission.
Size: 8.0 mm
Knife angle: 45°