Learn More
Sigma Aldrich Fine Chemicals Biosciences Histodenz, 66108-95-0, MFCD00077732, 100G
Catalog No. NC1589271
Encompass
Molecular Formula: C19H26I3N3O9, Molecular Weight: 821.14, Beilstein Registry Number: 2406632, nonionic density gradient medium
Catalog No. NC1589271 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D2158100G
Molecular Formula: C19H26I3N3O9, Molecular Weight: 821.14, Beilstein Registry Number: 2406632, nonionic density gradient medium