Learn More
Histodenz™, Sigma-Aldrich
Catalog No. 01259002 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
01-259-002 100 g
1 options
Catalog No. 01-259-002 Supplier MilliporeSigma Supelco Supplier No. D2158100G
Add to Cart
Add to Cart
HistoDenz™ is suitable for separating everything from macromolecules to viable cells by either rate-zonal or isopycnic centrifugation. It has been used in the isolation and culturing of hepatic stellate cells. It has also been employed in the extraction of microbial cells from environmental samples. It has been used in LacZ (B-galactosidase) staining. It has also been used for the enrichment of gonocytes.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Type Density Gradient Media
Form Powder