Learn More
StatLab HistoGel Biopsy Gel
SDP
Catalog No. 21010286
Change view
Click to view available options
Form:
Gel
Catalog No. Form
21-010-286 Gel
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-286 Supplier StatLab Supplier No. EMHGECS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Specimen processing gel for retaining small biopsies.
  • The simple way to retain small biopsies, FNA's, and cytology specimens for tissue processing and sectioning
  • No matter how small, friable, or viscous your histology or cytology specimen is, HistoGel will encapsulate and retain the entire specimen during histological processing
  • It allows fixatives, alcohols, clearing agents, and paraffins to infiltrate the specimen while processing
  • Sectioning is easy with HistoGel embedded specimens, allowing for perfect sections from scanty cellular material
  • The gel has no impact on histology or cytology stains
  • 1 case of twelve 10 mL tubes
Specifications
Product Type Specimen Processing Gel
Form Gel