Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Histology Grade 6.4 mm thick, 25 x 400 mm
SDP
Catalog No. 50930674
Encompass_Preferred
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 6.4 mm
Size: 25 x 400 mm
Catalog No. 50-930-674 Supplier Electron Microscopy Sciences Supplier No. 71016
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 6.4 mm
Size: 25 x 400 mm