Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Histology Brush with Magnetic End
SDP

Catalog No. 22000085
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-000-085 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-085 Supplier StatLab Supplier No. TSDB
Add to Cart
Add to Cart

Ox hair tissue section debris brush is used to aid microtomy and ensure the clean working area of the microtome when sectioning

  • The magnetic end helps to promote safety and avoid potential injury by eliminating the need for techs to use their hands to remove microtome blades
  • 9 in.

Specifications

Product Type Histology Brush
For Use With (Equipment) Microtome
Material Wood and Ox Hair
Disposable Reusable
Length (English) 9 in.
Length (Metric) 228.6 mm
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.