Learn More
Volu Sol Saccomanno Solution, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VMA128
Encompass_Preferred
Premixed fixative, used in collecting mucoid specimens for cytologic diagnosis
Catalog No. 50-420-861
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.