Learn More
Azer Scientific Histology Tissue Control Slides
SDP
Catalog No. 22026733
Change view
Click to view available options
Product Type:
Alcian Blue Standard
Amyloid Standard
Argentaffin Standard
ER Immunohistochemistry Control Slide
Elastic Tissue Standard
Fungi-GMS Standard
Fungi-PAS Standard
Giemsa Standard
Gram Organism Standard
HER-2Nu
HPV
Iron Standard Histology Control Slide
Luxol Fast Blue Standard
Massons Standard
Melanin Standard
Mucicarmine Standard
Normal Cervix
PIN-4
PR Immunohistochemistry Control Slide
PSA
PTAH Standard Histology Control Slide
Periodic Acid Standard
Pneumocystis Standard
Reticulum Standard
Trichrome Standard
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-026-733 HPV
22-026-718 Melanin Standard
22-026-725 PIN-4
22-026-732 Normal Cervix
22-026-705 Argentaffin Standard
22-026-728 ER Immunohistochemistry Control Slide
22-026-727 PSA
22-026-721 PTAH Standard Histology Control Slide
22-026-729 PR Immunohistochemistry Control Slide
22-026-719 Mucicarmine Standard
22-026-717 Massons Standard
22-026-720 Periodic Acid Standard
22-026-716 Luxol Fast Blue Standard
22-026-708 Elastic Tissue Standard
22-026-722 Reticulum Standard
22-026-702 Pneumocystis Standard
22-026-715 Iron Standard Histology Control Slide
22-026-704 Amyloid Standard
22-026-714 Gram Organism Standard
22-026-730 HER-2Nu
22-026-723 Trichrome Standard
22-026-703 Alcian Blue Standard
22-026-710 Fungi-PAS Standard
22-026-711 Fungi-GMS Standard
22-026-712 Giemsa Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. 22-026-733 Supplier Azer Scientific Supplier No. ESCS180
Add to Cart
Add to Cart
Includes
10 unstained and 1 stained Unifrost slide
Designed for use in monitoring stain performance in the pathology laboratory
  • Prepared with paraffin-embedded human tissue sections
  • For QC of laboratory's staining processes
Specifications
For Use With (Application) Designed for use in monitoring stain performance in the pathology laboratory
Includes 10 unstained and 1 stained unifrost slide
Quantity 11 Pack
Product Type HPV