Learn More
Laxco™ Histology Upgrade Kit for SeBaP4C2
GREENER_CHOICESDP
Catalog No. 12563680 Shop All Laxco Inc Products
Change view
Click to view available options
For Use With (Equipment):
SEBAP4C2 Microscope
Catalog No. For Use With (Equipment)
12-563-680 SEBAP4C2 Microscope
1 options
Catalog No. 12-563-680 Supplier Laxco™ Supplier No. SEBAP4C2HISTKIT
Add to Cart
Add to Cart
Histology upgrade kit for analyzing blood (hematology) samples, various histological sample slides and diagnosis.
Advanced histology kit.
  • Your go-to kit for all histological analysis and imaging needs
  • Research grade PLAN achromat 50x oil immersion objective
Specifications
Type Adapter Kit
Autoclavable Not Autoclavable
For Use With (Equipment) SEBAP4C2 Microscope
Sterility Sterile
Voltage 100/115/220/230 V