Learn More
Seracare Life Sciences Inc HISTOMARK PHTHALORED SOLUTION
Supplier:  Seracare Life Sciences Inc 55100038
Encompass
PhThaloRED Solution for Immunohistochemistry is a New Fuchsin stain. It is a component of the HistoMark RED Substrate System for use with alkaline phosphatase. Requires Activator Solution and Buffered Substrate Solution for use.
Catalog No. 50-675-52