Learn More
Seracare Life Sciences Inc HISTOMARK X-GAL SUBSTRATE SET
Catalog No. 5067502
Encompass_Preferred
The HistoMark X-Gal Substrate Set is designed for use with any probe labeled with E. coli B-galactosidase. The substrate produces a brilliant blue specific stain with no background color. The stain is permanent and insoluble in alcohols or xylene.
This item is not returnable. View return policy
Catalog No. 50-675-02 Supplier Seracare Life Sciences Inc Supplier No. 55200022
The HistoMark X-Gal Substrate Set is designed for use with any probe labeled with E. coli B-galactosidase. The substrate produces a brilliant blue specific stain with no background color. The stain is permanent and insoluble in alcohols or xylene.