Learn More
Seracare Life Sciences Inc HISTOMARKBLACK HRP SUBS KIT
Catalog No. 5067517
Encompass_Preferred
Metal enhanced DAB technology permits detection of extremely low concetrations of tissue antigens. This kit provides a sensitive means for detecting peroxidase labeled antibodies and conjugates. HistoMark Black contains cobalt-enhanced DAB that produces a black, cobalt- DAB polymer with a signal 5-10 times more sensitive than DAB alone. The kit contains the nuclear counterstain, Contrast GREEN, a solution of methyl green that contrasts sharply with black. Blocking solution is provided to prevent endogenous peroxidase activity. The precipitates from HistoMark Black are alcohol and xylene insoluble, allowing section to be mounted in aqueous or xylene-based mounting media.
This item is not returnable. View return policy
Catalog No. 50-675-17 Supplier Seracare Life Sciences Inc Supplier No. 55100034
Metal enhanced DAB technology permits detection of extremely low concetrations of tissue antigens. This kit provides a sensitive means for detecting peroxidase labeled antibodies and conjugates. HistoMark Black contains cobalt-enhanced DAB that produces a black, cobalt- DAB polymer with a signal 5-10 times more sensitive than DAB alone. The kit contains the nuclear counterstain, Contrast GREEN, a solution of methyl green that contrasts sharply with black. Blocking solution is provided to prevent endogenous peroxidase activity. The precipitates from HistoMark Black are alcohol and xylene insoluble, allowing section to be mounted in aqueous or xylene-based mounting media.