Learn More
Cayman Chemical HIston H1-derIved PeptIdet 1mg
SDP
Catalog No. 502941040
Encompass_Preferred
A peptide substrate for CRK3/CYC6; phosphorylated by CRK3/CYC6 and has been used in high-throughput screening assays for the identification of CRK3/CYC6 inhibitors
Catalog No. 50-294-1040 Supplier Cayman Chemical Supplier No. 274391 MG
A peptide substrate for CRK3/CYC6; phosphorylated by CRK3/CYC6 and has been used in high-throughput screening assays for the identification of CRK3/CYC6 inhibitors