Learn More
Enzo Life Sciences Histone H1 Recombinant monoclonal antibody (HH1/1784R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3750100
Encompass_Preferred
Clone: HH1/1784R. Isotype: IgGκ. Immunogen: Recombinant full length human Histone H1. Species reactivity: Human, Mouse, Rat. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2902