Learn More
Cayman Chemical HIston H2AXC-Term Monclo 100ug
SDP
Catalog No. 502948268
Encompass_Preferred
Immunogen: Peptide from the C-terminal region of human H2AX • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H2AX independent of PTMs; (-) Other histone proteins • Applications: ELISA, ICC, multiplex-based assays, WB
Catalog No. 50-294-8268 Supplier Cayman Chemical Supplier No. 32183100 UG
Immunogen: Peptide from the C-terminal region of human H2AX • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H2AX independent of PTMs; (-) Other histone proteins • Applications: ELISA, ICC, multiplex-based assays, WB