Learn More
Cayman Chemical HIston H2BC-Term Monclon 100ug
SDP
Catalog No. 502948083
Encompass_Preferred
Immunogen: Peptide corresponding to the C-terminal region of human H2B • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H2B independent of PTMs; (-) H2A, other histone proteins • Applications: ELISA, ICC, multiplex-based assays, WB
Catalog No. 50-294-8083 Supplier Cayman Chemical Supplier No. 32182100 UG
Immunogen: Peptide corresponding to the C-terminal region of human H2B • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H2B independent of PTMs; (-) H2A, other histone proteins • Applications: ELISA, ICC, multiplex-based assays, WB