Learn More
Cayman Chemical HIston H321-44-GK-bIotIn 250ug
SDP
Supplier:  Cayman Chemical 27761250 UG
Encompass_Preferred
A biotinylated peptide fragment of histone H3; has been used as a substrate for PRDM7 to determine substrate specificity
Catalog No. 50-293-8861