Learn More
Cayman Chemical HIston H3CItrulInated R2 100ug
Supplier: Cayman Chemical 17939100 UG
Immunogen: Histone H3 peptide with citrulline at R2, R8, and R17 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (-) Non-citrullinated histone H3 • Applications: ELISA, WB • MW = ~16 kDa