Learn More
Cayman Chemical HIston H3CItrulInated R2 100ug
SDP
Catalog No. 502942194
Encompass_Preferred
Preparation: This antibody, which contains a mouse IgG isoform, was produced as a recombinant protein expressed in HEK293-derived cells using an engineered construct developed based on the Histone H3 (Citrullinated R2 + R8 + R17) Monoclonal Antibody (Clone 11D3) • Host: HEK293-derived cells • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) Citrullinated histone H3 (-) Histone H3, other citrullinated proteins • Applications: ELISA, WB
Catalog No. 50-294-2194 Supplier Cayman Chemical Supplier No. 40359100 UG
Preparation: This antibody, which contains a mouse IgG isoform, was produced as a recombinant protein expressed in HEK293-derived cells using an engineered construct developed based on the Histone H3 (Citrullinated R2 + R8 + R17) Monoclonal Antibody (Clone 11D3) • Host: HEK293-derived cells • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) Citrullinated histone H3 (-) Histone H3, other citrullinated proteins • Applications: ELISA, WB