Learn More
Cayman Chemical Histone H3 (Citrullinated R2 + R8 + R17) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502968182
Encompass_Preferred
Immunogen: Histone H3 peptide with citrullinations at R2, R8, and R17 • Host: Rabbit • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (-) Non-citrullinated histone H3 • Applications: ELISA, WB • MW = 16 kDa
Catalog No. 50-296-8182 Supplier Cayman Chemical Supplier No. 178551 EA
Immunogen: Histone H3 peptide with citrullinations at R2, R8, and R17 • Host: Rabbit • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (-) Non-citrullinated histone H3 • Applications: ELISA, WB • MW = 16 kDa