Learn More
Medchemexpress LLC HY-100734 5mg Medchemexpress, Histone Acetyltransferase Inhibitor II CAS:932749-62-7 Purity:>98%
SDP
Catalog No. 501931930
Encompass_Preferred
Medchemexpress, HY-100734 5mg Histone Acetyltransferase Inhibitor II CAS:932749-62-7 Histone Acetyltransferase Inhibitor II is a potent and cell permeable p300 inhibitor, with an IC 50 of 5 µM; Histone Acetyltransferase Inhibitor II can be used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1930 Supplier Medchemexpress LLC Supplier No. HY1007345MG
Medchemexpress, HY-100734 5mg Histone Acetyltransferase Inhibitor II CAS:932749-62-7 Histone Acetyltransferase Inhibitor II is a potent and cell permeable p300 inhibitor, with an IC 50 of 5 µM; Histone Acetyltransferase Inhibitor II can be used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.