Learn More
TARGETMOL CHEMICALS INC Histone Acetyltransferase Inhi
SDP
Catalog No. 502880787
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Histone Acetyltransferase Inhibitor II is a selective and cell permeable inhibitor of p300 histone acetyltransferase(IC50 5 uM).with anti-acetylase activity in mammalian cells. Purity 97.13%
Catalog No. 50-288-0787 Supplier TARGETMOL CHEMICALS INC Supplier No. T1156325MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Histone Acetyltransferase Inhibitor II is a selective and cell permeable inhibitor of p300 histone acetyltransferase(IC50 5 uM).with anti-acetylase activity in mammalian cells. Purity 97.13%