Learn More
Active Motif HAT Assay Kit (Fluorescent)
SDP
Catalog No. NC0008851
Encompass
The HAT Assay Kit is a fluorescent method to assay samples for histone acetyltransferase (HAT) activity or to screen HAT inhibitors. The assay is easy to use, features a simple protocol that gives you results in 30 minutes and is ideal for high-throughput usage in 96-well plates.
Catalog No. NC0008851 Supplier Active Motif Supplier No. 56100
The HAT Assay Kit is a fluorescent method to assay samples for histone acetyltransferase (HAT) activity or to screen HAT inhibitors. The assay is easy to use, features a simple protocol that gives you results in 30 minutes and is ideal for high-throughput usage in 96-well plates.