Learn More
Medchemexpress LLC Histone deacetylase 100ug
SDP
Catalog No. 5000181354
Encompass_Preferred
Studies have confirmed that the histone deacetylase 1 (HDAC1) protein is a key enzyme that deacetylates lysine residues on core histones (H2A H2B H3 H4) This process establishes an epigenetic repressive signature that affects transcription cell cycle and developmental events Histone deacetylase 1/HDAC1 Protein Human (His-SUMO) is the recombinant human-derived Histone deacetylase 1/HDAC1 protein expressed by E coli with N-SUMO N-6 His labeled tag
Catalog No. 50-001-81354 Supplier Medchemexpress LLC Supplier No. HYP72262100UG
Studies have confirmed that the histone deacetylase 1 (HDAC1) protein is a key enzyme that deacetylates lysine residues on core histones (H2A H2B H3 H4) This process establishes an epigenetic repressive signature that affects transcription cell cycle and developmental events Histone deacetylase 1/HDAC1 Protein Human (His-SUMO) is the recombinant human-derived Histone deacetylase 1/HDAC1 protein expressed by E coli with N-SUMO N-6 His labeled tag