Learn More
Arbor Assays HDM Fluorescent Kit (2 Plate)
SDP
Catalog No. 502278392
Encompass_Preferred
The DetectX Universal Histone Demethylase (HDM) Fluorescent Activity Kit allows activity measurement of any Histone Demethylase system by measuring the formaldehyde generated by demethylation in low, medium and high throughput fashion.
This item is not returnable. View return policy
Catalog No. 50-227-8392 Supplier Arbor Assays Supplier No. K010F1
The DetectX Universal Histone Demethylase (HDM) Fluorescent Activity Kit allows activity measurement of any Histone Demethylase system by measuring the formaldehyde generated by demethylation in low, medium and high throughput fashion.