Learn More
Enzo Life Sciences EPIXTRACT Total Histone Extraction Kit (High Throughput) (96Reactions)
SDP
Catalog No. 502012571
Encompass_Preferred
EPIXTRACT Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays. Assay Time: 1 hour.
Catalog No. 50-201-2571 Supplier Enzo Life Sciences Supplier No. ENZ450130002
EPIXTRACT Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays. Assay Time: 1 hour.