Learn More
Sigma Aldrich Fine Chemicals Biosciences Histone from calf thymus
Catalog No. 502905763
Encompass
Mixture of histones H1 H2A H2B H3 and H4 isolated from calf thymus.Histones are a group of DNA-binding proteins that are characterized by relatively high levels of lysine and arginine. five different fractions of histones have been isolated and characterized. These are named as H1 H2A H2B H3 and H4. The H1 fraction is lysine-rich while the H2A and H2B are slightly lysine-rich The H3 and H4 fractions are arginine-rich. The molecular weights of histones are approximately 11 to 21 kDa depending on the fraction.
Catalog No. 50-290-5763 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10223565001
Mixture of histones H1 H2A H2B H3 and H4 isolated from calf thymus.Histones are a group of DNA-binding proteins that are characterized by relatively high levels of lysine and arginine. five different fractions of histones have been isolated and characterized. These are named as H1 H2A H2B H3 and H4. The H1 fraction is lysine-rich while the H2A and H2B are slightly lysine-rich The H3 and H4 fractions are arginine-rich. The molecular weights of histones are approximately 11 to 21 kDa depending on the fraction.