Learn More
Medchemexpress LLC Histone H1 (calf thymus) | 1523001-32-2 | 25 MG
SDP
Catalog No. 5000365774
Encompass_Preferred
Histone H1 (calf thymus) is a linker histone subtype that is found in the deoxyribonucleoprotein of calf thymus and can be obtained by separation and purification from total calf thymus histones. For research use only.
  • Obtained by separation and purification from total calf thymus histones
  • For research use only
Catalog No. 50-003-65774 Supplier Medchemexpress LLC Supplier No. 25MGHYNP155A
Histone H1 (calf thymus) is a linker histone subtype that is found in the deoxyribonucleoprotein of calf thymus and can be obtained by separation and purification from total calf thymus histones. For research use only.
  • Obtained by separation and purification from total calf thymus histones
  • For research use only