Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Histone H1 (calf thymus) | 1523001-32-2 | 91% | 10 MG
SDP

Catalog No. 5000365525
Encompass_Preferred

Histone H1 (calf thymus) is a linker histone subtype found in the deoxyribonucleoprotein of calf thymus. It is obtained by separation and purification from total calf thymus histones.

  • White to off-white solid appearance.
  • Derived from bovine (calf) thymus.
  • Purity of 91% (protein by Biuret).
  • Suitable for research use only.
  • Can be stored at -20°C for up to 2 years.

Catalog No. 50-003-65525 Supplier Medchemexpress LLC Supplier No. 10MGHYNP155A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Histone H1 (calf thymus) is a linker histone subtype found in the deoxyribonucleoprotein of calf thymus. It is obtained by separation and purification from total calf thymus histones.

  • White to off-white solid appearance.
  • Derived from bovine (calf) thymus.
  • Purity of 91% (protein by Biuret).
  • Suitable for research use only.
  • Can be stored at -20°C for up to 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.