Learn More
Medchemexpress LLC Histone H1 (calf thymus) | 1523001-32-2 | 91% | 100 MG
SDP
Catalog No. 5000365004
Encompass_Preferred
Histone H1 (calf thymus) is a linker histone subtype found in the deoxyribonucleoprotein of calf thymus. It is obtained by separation and purification from total calf thymus histones.
  • Linker histone subtype
  • Derived from calf thymus deoxyribonucleoprotein
  • Obtained by separation and purification from total calf thymus histones
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-65004 Supplier Medchemexpress LLC Supplier No. 100MGHYNP155A
Histone H1 (calf thymus) is a linker histone subtype found in the deoxyribonucleoprotein of calf thymus. It is obtained by separation and purification from total calf thymus histones.
  • Linker histone subtype
  • Derived from calf thymus deoxyribonucleoprotein
  • Obtained by separation and purification from total calf thymus histones
  • For research use only
  • Appearance: Solid
  • Color: White to off-white