Learn More
Medchemexpress LLC Histone H1-derived Peptide | 889112-06-5 | 99.9% | 1252.51 | 10 MG
SDP
Catalog No. 5000423894
Encompass_Preferred
Histone H1-derived Peptide is a phosphopeptide whose peptide substrates contain a sequence in accordance with the optimal recognition motif for CDK. It can be used for detecting CDK1-cyclinB1 enzyme activity.
  • Phosphopeptide designed for CDK activity detection
  • Contains sequence matching optimal recognition motif for CDK
  • Used for detecting CDK1-cyclinB1 enzyme activity
Catalog No. 50-004-23894 Supplier Medchemexpress LLC Supplier No. HYP248010MG
Histone H1-derived Peptide is a phosphopeptide whose peptide substrates contain a sequence in accordance with the optimal recognition motif for CDK. It can be used for detecting CDK1-cyclinB1 enzyme activity.
  • Phosphopeptide designed for CDK activity detection
  • Contains sequence matching optimal recognition motif for CDK
  • Used for detecting CDK1-cyclinB1 enzyme activity