Learn More
CPC Scientific 5TAMRA-Gly-Gly-Gly-Pro-Ala-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIST003A
Encompass
A TamrA-labeled histone H1-derived peptide (Ab/Em = 544/572 nm) that is phosphorylated by protein kinase A and serves as a substrate for CDK2 and CDK5 (cyclin dependent kinase 5) and PKA. ONE-LETTER SEQUENCE: 5TAMRA-GGGPATPKKAKKLMOLECULAR FORMULA: C81H121N19O19MOLECULAR WEIGHT:1665STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Pomerant, AM. et al. Proc. Natl. Acad. Sci. USA. 74, 4261 (1977)
Catalog No. 50-210-9426