Learn More
Sino Biological Peptide Substrates: Histone H1 Peptide
Catalog No. 502569047
Encompass_Preferred
The Histone H1 Peptide (152-159) sequence (GGGPATP-KKAKKL-COOH) is derived from human histone H1 between amino acids 152-159 and is suitable for use as the substrate for CDK family kinase assays, such as CDK1, CDK2, CDK5 and CDK6.
Catalog No. 50-256-9047 Supplier Sino Biological Supplier No. H10581
The Histone H1 Peptide (152-159) sequence (GGGPATP-KKAKKL-COOH) is derived from human histone H1 between amino acids 152-159 and is suitable for use as the substrate for CDK family kinase assays, such as CDK1, CDK2, CDK5 and CDK6.