Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Histone H1 protein, human (His) | 1523001-32-2 | >90.0% | 20 UG
SDP

Catalog No. 5000291243
Encompass_Preferred

Recombinant human histone H1 (H1.0 variant) with an N-terminal His tag, expressed in Escherichia coli and supplied as a filtered, stabilized solution for research use in chromatin, nucleosome, and epigenetics studies.

  • Recombinant full-length human H1.0 with N-terminal His tag.
  • Expressed in Escherichia coli for consistent production.
  • Supplied as a 0.22 μm filtered solution with stabilizers (trehalose, mannitol, DTT, Tween 80).
  • Purity >90.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg for sensitive biological assays.
  • Stable when stored frozen; shipped on dry ice to preserve activity.

Catalog No. 50-002-91243 Supplier Medchemexpress LLC Supplier No. HYP7488720UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human histone H1 (H1.0 variant) with an N-terminal His tag, expressed in Escherichia coli and supplied as a filtered, stabilized solution for research use in chromatin, nucleosome, and epigenetics studies.

  • Recombinant full-length human H1.0 with N-terminal His tag.
  • Expressed in Escherichia coli for consistent production.
  • Supplied as a 0.22 μm filtered solution with stabilizers (trehalose, mannitol, DTT, Tween 80).
  • Purity >90.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg for sensitive biological assays.
  • Stable when stored frozen; shipped on dry ice to preserve activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.