Learn More
Apexbio Technology LLC Histone-H2A-(107-122)-Ac-OH 25mg
SDP
Catalog No. 5000093079
Encompass_Preferred
Histone-H2A-107-122-Ac-OH is a synthetic peptide derived from the C-terminal region of the histone H2A protein It is designed as a molecular tool to investigate chromatin-associated processes and histone modifications thereby facilitating the study of epigenetic regulatory mechanisms and gene expression control Histone-H2A-107-122-Ac-OH exerts its biological activity primarily through mimicking a histone H2A segment enabling research into the roles of this core histone in nucleosome assembly DNA compaction and chromatin remodeling Based on these properties Histone-H2A-107-122-Ac-OH holds research potential in studying chromatin dynamics epigenetic regulation and related biomedical applications
Catalog No. 50-000-93079 Supplier Apexbio Technology LLC Supplier No. A114625
Histone-H2A-107-122-Ac-OH is a synthetic peptide derived from the C-terminal region of the histone H2A protein It is designed as a molecular tool to investigate chromatin-associated processes and histone modifications thereby facilitating the study of epigenetic regulatory mechanisms and gene expression control Histone-H2A-107-122-Ac-OH exerts its biological activity primarily through mimicking a histone H2A segment enabling research into the roles of this core histone in nucleosome assembly DNA compaction and chromatin remodeling Based on these properties Histone-H2A-107-122-Ac-OH holds research potential in studying chromatin dynamics epigenetic regulation and related biomedical applications