Learn More
Abcam Rabbit polyclonal to Histone H2A.X - Nuclear MarkerSuitable for: WB, IHC-PKnockout validatedReacts with: Cow, HumanIsotype: IgG
Supplier:  Abcam AB10475100UG
Encompass_Preferred
Rabbit polyclonal to Histone H2A.X - Nuclear MarkerSuitable for: WB, IHC-PKnockout validatedReacts with: Cow, HumanIsotype: IgG
Catalog No. NC2467396