Learn More
Active Motif Histone H2AK119ub antibody (pAb)
SDP
Supplier:  Active Motif 65697
Encompass
Active Motif's Histone H2AK119ub antibody (pAb) was raised in a Rabbit host. It has been validated for use in Dot blot and Western blot; it has been shown to react with Human samples, but it is predicted that it will react with a wide range of sample types. For more complete and up-to-date information, please visit the Active Motif website.
Catalog No. NC1850694